Skip to Content
Merck
All Photos(1)

Key Documents

8.06754

Sigma-Aldrich

Ethyl 3-pyridinecarboxylate

for synthesis

Synonym(s):

Ethyl 3-pyridinecarboxylate, Ethyl nicotinate, 3-Pyridinecarboxylic acid ethyl ester, Nicotinic acid ethyl ester

Sign Into View Organizational & Contract Pricing

Select a Size

250 ML
€112.00

€112.00


Estimated to ship on07 May 2025Details


Request a Bulk Order

Select a Size

Change View
250 ML
€112.00

About This Item

Linear Formula:
3-(C2H5OOC)C5H4N
CAS Number:
Molecular Weight:
151.16
MDL number:
UNSPSC Code:
12352108
EC Index Number:
210-370-7
NACRES:
NA.22

€112.00


Estimated to ship on07 May 2025Details


Request a Bulk Order

Quality Level

Assay

≥98.0% (GC)

form

liquid

potency

>2005 mg/kg LD50, oral (Rat)

bp

222-224 °C/1013 mbar

mp

8-9 °C

transition temp

flash point 93 °C

density

1.11 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

n1cc(ccc1)C(=O)OCC

InChI

1S/C8H9NO2/c1-2-11-8(10)7-4-3-5-9-6-7/h3-6H,2H2,1H3

InChI key

XBLVHTDFJBKJLG-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.106 - 1.108
Identity (IR): passes test

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible, corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

199.4 °F - closed cup

Flash Point(C)

93 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service