Skip to Content
Merck
All Photos(1)

Key Documents

8.05807

Sigma-Aldrich

3-Methylcyclohexanol

(mixture of cis- and trans isomers) for synthesis

Synonym(s):

3-Methylcyclohexanol

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
€177.00

€177.00


Estimated to ship on31 March 2025Details



Select a Size

Change View
100 ML
€177.00

About This Item

Empirical Formula (Hill Notation):
C7H14O
CAS Number:
Molecular Weight:
114.19
MDL number:
UNSPSC Code:
12352104
EC Index Number:
209-709-1
NACRES:
NA.22

€177.00


Estimated to ship on31 March 2025Details


vapor pressure

1.7 hPa ( 25 °C)

Quality Level

Assay

≥98.0% (GC)

autoignition temp.

295 °C

expl. lim.

1.3 % (v/v)

mp

-41 °C

transition temp

flash point 70 °C

density

0.92 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

OC1CC(CCC1)C

InChI

1S/C7H14O/c1-6-3-2-4-7(8)5-6/h6-8H,2-5H2,1H3

InChI key

HTSABYAWKQAHBT-UHFFFAOYSA-N

General description

3-Methylcyclohexanol is a cyclic secondary alcohol serves primarily as an intermediate in the synthesis of ketones, such as 3-methylcyclohexanone, a precursor for key compounds like isophorone. Its reactivity allows it to undergo transformations like oxidation, dehydration, and substitution, enabling the synthesis of diverse compounds.

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.916 - 0.918
Identity (IR): passes test

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Inhalation

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

145.4 °F - closed cup

Flash Point(C)

63 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Dye-sensitized solar cells (DSCs) are 3rd generation solar cells combining the promise of high efficiency with low production costs.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service