Skip to Content
Merck
All Photos(1)

Key Documents

8.05805

Sigma-Aldrich

Quinaldine

for synthesis

Synonym(s):

Quinaldine, 2-Methylquinoline

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
€84.60
250 ML
€138.00

€84.60


Estimated to ship on14 April 2025Details


Request a Bulk Order

Select a Size

Change View
100 ML
€84.60
250 ML
€138.00

About This Item

Empirical Formula (Hill Notation):
C10H9N
CAS Number:
Molecular Weight:
143.19
MDL number:
UNSPSC Code:
12352005
EC Index Number:
202-085-1
NACRES:
NA.22

€84.60


Estimated to ship on14 April 2025Details


Request a Bulk Order

vapor pressure

<0.1 hPa ( 20 °C)

Quality Level

Assay

≥98.0% (GC)

form

liquid

potency

1230 mg/kg LD50, oral (Rat)
1870 mg/kg LD50, skin (Rabbit)

pH

6.9 (20 °C in H2O, saturated aqueous solution)

bp

246.9 °C/1013 hPa

mp

-2 °C

transition temp

flash point 79 °C

density

1.06 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

n1c2c(ccc1C)cccc2

InChI

1S/C10H9N/c1-8-6-7-9-4-2-3-5-10(9)11-8/h2-7H,1H3

InChI key

SMUQFGGVLNAIOZ-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.059 - 1.061
Identity (IR): passes test

Pictograms

Exclamation markHealth hazard

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Irrit. 2 - Muta. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

174.2 °F - closed cup

Flash Point(C)

79 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service