Skip to Content
Merck
All Photos(1)

Key Documents

8.04789

Sigma-Aldrich

2-Imidazolidinone hemihydrate

for synthesis

Synonym(s):

2-Imidazolidinone hemihydrate, N,N′-Ethyleneurea

Sign Into View Organizational & Contract Pricing

Select a Size

500 G
€53.00
50 KG
€1,950.00

€53.00


Estimated to ship on27 March 2025Details


Request a Bulk Order

Select a Size

Change View
500 G
€53.00
50 KG
€1,950.00

About This Item

Empirical Formula (Hill Notation):
C3H6N2O · 0.5H2O
CAS Number:
Molecular Weight:
95.10
MDL number:
UNSPSC Code:
12352115
EC Index Number:
204-436-4
NACRES:
NA.22

€53.00


Estimated to ship on27 March 2025Details


Request a Bulk Order

vapor pressure

<24 hPa ( 20 °C)

Quality Level

form

crystals

autoignition temp.

420 °C

potency

>5000 mg/kg LD50, oral (Rat)

pH

9-11 (20 °C, 100 g/L in H2O)

bp

335.5 °C/1013.25 hPa (anhydrous substance)

mp

58 °C

transition temp

flash point 265 °C

density

1.3 g/cm3 at 20 °C

bulk density

720 kg/m3

storage temp.

2-30°C

SMILES string

N1CCNC1=O

InChI

1S/C3H6N2O/c6-3-4-1-2-5-3/h1-2H2,(H2,4,5,6)

InChI key

YAMHXTCMCPHKLN-UHFFFAOYSA-N

Analysis Note

Assay (ex N): ≥ 97.0 %
Water (K. F.): ≤ 13.0 %
Identity (IR): passes test

Pictograms

Exclamation markHealth hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - STOT RE 2

Target Organs

Thyroid

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

509.0 °F

Flash Point(C)

265 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service