Skip to Content
Merck
All Photos(1)

Key Documents

8.03501

Sigma-Aldrich

1,12-Dodecanediol

for synthesis

Synonym(s):

1,12-Dodecanediol, 1,12-Dihydroxydodecane, Dodecamethylene glycol

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
€93.90

€93.90


Estimated to ship on31 March 2025Details


Request a Bulk Order

Select a Size

Change View
100 G
€93.90

About This Item

Linear Formula:
HO(CH2)12OH
CAS Number:
Molecular Weight:
202.33
MDL number:
UNSPSC Code:
12352104
EC Index Number:
227-133-9
NACRES:
NA.22

€93.90


Estimated to ship on31 March 2025Details


Request a Bulk Order

Quality Level

Assay

≥97.5% (GC)

form

crystals

autoignition temp.

300 °C

bp

324 °C/1013 hPa

mp

79 °C

transition temp

flash point 176 °C

storage temp.

2-30°C

SMILES string

OCCCCCCCCCCCCO

InChI

1S/C12H26O2/c13-11-9-7-5-3-1-2-4-6-8-10-12-14/h13-14H,1-12H2

InChI key

GHLKSLMMWAKNBM-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 97.5 % (a/a)
Melting range (lower value): ≥ 78 °C
Melting range (upper value): ≤ 82 °C
Identity (IR): passes test

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

348.8 °F - closed cup

Flash Point(C)

176.00 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service