Skip to Content
Merck
All Photos(1)

Key Documents

8.03264

Sigma-Aldrich

1,1-Iminodi-2-propanol

for synthesis

Synonym(s):

1,1-Iminodi-2-propanol, Diisopropanolamine, Bis(2-hydroxypropyl)amine

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
€33.40
1 KG
€63.80
25 KG
€490.00

€33.40


Estimated to ship on02 May 2025Details


Request a Bulk Order

Select a Size

Change View
100 G
€33.40
1 KG
€63.80
25 KG
€490.00

About This Item

Linear Formula:
[CH3CH(OH)CH2]2NH
CAS Number:
Molecular Weight:
133.19
MDL number:
UNSPSC Code:
12352104
EC Index Number:
203-820-9
NACRES:
NA.22

€33.40


Estimated to ship on02 May 2025Details


Request a Bulk Order

vapor pressure

0.02 hPa ( 20 °C)

Quality Level

Assay

≥98.0% (GC)

form

solid

autoignition temp.

290 °C

potency

4765 mg/kg LD50, oral (Rat)
8000 mg/kg LD50, skin (Rabbit)

expl. lim.

1.6-8.0 % (v/v)

pH

11.4 (20 °C, 100 g/L in H2O)

bp

249 °C/1013 hPa

mp

40.0-48.0 °C

transition temp

flash point 135 °C

solubility

870 g/L

density

0.99 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

N(CC(O)C)CC(O)C

InChI

1S/C6H15NO2/c1-5(8)3-7-4-6(2)9/h5-9H,3-4H2,1-2H3

InChI key

LVTYICIALWPMFW-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Water (according to Karl Fischer): ≤ 1.0 %
Identity (IR): passes test
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

275.0 °F - closed cup

Flash Point(C)

135 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service