Skip to Content
Merck
All Photos(2)

Key Documents

8.03247

Sigma-Aldrich

1-(Dimethylamino)-2-propanol

for synthesis

Synonym(s):

1-(Dimethylamino)-2-propanol, N-(2-Hydroxypropyl)-N,N-dimethylamine, N,N-Dimethylisopropanolamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH(OH)CH2N(CH3)2
CAS Number:
Molecular Weight:
103.16
MDL number:
UNSPSC Code:
12352104
EC Index Number:
203-556-4
NACRES:
NA.22
Pricing and availability is not currently available.

vapor pressure

17.5 hPa ( 20 °C)

Quality Level

Assay

≥99.0% (GC)

form

liquid

autoignition temp.

225 °C

potency

1360 mg/kg LD50, oral (Rat)

expl. lim.

2.7-11.1 % (v/v)

pH

11 (20 °C, 100 g/L in H2O)

bp

125-127 °C/1013 hPa

mp

-85 °C

transition temp

flash point 26 °C

density

0.85 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

N(CC(O)C)(C)C

InChI

1S/C5H13NO/c1-5(7)4-6(2)3/h5,7H,4H2,1-3H3

InChI key

NCXUNZWLEYGQAH-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.849 - 0.850
Identity (IR): passes test

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

78.8 °F - DIN 51755 Part 1

Flash Point(C)

26 °C - DIN 51755 Part 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service