Skip to Content
Merck
All Photos(1)

Key Documents

8.03214

Sigma-Aldrich

N,N-Diethylacetamide

for synthesis

Synonym(s):

N,N-Diethylacetamide, Acetic acid diethylamide

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
€89.50

€89.50


Estimated to ship on20 March 2025Details


Request a Bulk Order

Select a Size

Change View
100 ML
€89.50

About This Item

Linear Formula:
CH3CON(C2H5)2
CAS Number:
Molecular Weight:
115.17
MDL number:
UNSPSC Code:
12352111
EC Index Number:
211-685-2
NACRES:
NA.22

€89.50


Estimated to ship on20 March 2025Details


Request a Bulk Order

vapor pressure

1.2 hPa ( 25 °C)

Quality Level

Assay

≥98.0% (GC)

form

liquid

potency

1500 mg/kg LD50, oral (Rat)

bp

182-186 °C/1013 hPa

transition temp

flash point 76 °C

density

0.91 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

N(CC)(CC)C(=O)C

InChI

1S/C6H13NO/c1-4-7(5-2)6(3)8/h4-5H2,1-3H3

InChI key

AJFDBNQQDYLMJN-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.907 - 0.911
Water (K. F.): ≤ 0.30 %
Identity (IR): passes test

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

168.8 °F - closed cup

Flash Point(C)

76 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service