Skip to Content
Merck
All Photos(1)

Key Documents

8.03120

Sigma-Aldrich

1,4-Dinitrobenzene

for synthesis

Synonym(s):

1,4-Dinitrobenzene

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
€83.90

€83.90


Estimated to ship on02 April 2025Details



Select a Size

Change View
10 G
€83.90

About This Item

Linear Formula:
1,4-(NO2)2C6H4
CAS Number:
Molecular Weight:
168.11
MDL number:
UNSPSC Code:
12352102
EC Index Number:
202-833-7
NACRES:
NA.22

€83.90


Estimated to ship on02 April 2025Details


Quality Level

Assay

≥94.0% (HPLC)

bp

299 °C/1013 hPa

mp

174 °C

transition temp

flash point 150 °C

solubility

0.069 g/L

storage temp.

2-30°C

SMILES string

[N+](=O)([O-])c1ccc(cc1)[N+](=O)[O-]

InChI

1S/C6H4N2O4/c9-7(10)5-1-2-6(4-3-5)8(11)12/h1-4H

InChI key

FYFDQJRXFWGIBS-UHFFFAOYSA-N

Analysis Note

Assay (HPLC, area%): ≥ 94.0 % (a/a)
Identity (IR): passes test

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 1 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2

Storage Class Code

6.1A - Combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

302.0 °F - closed cup

Flash Point(C)

150 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service