Skip to Content
Merck
All Photos(1)

Key Documents

8.02971

Sigma-Aldrich

3,4-Dihydro-2H-pyran

for synthesis

Synonym(s):

3,4-Dihydro-2H-pyran

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H8O
CAS Number:
Molecular Weight:
84.12
MDL number:
UNSPSC Code:
12352005
EC Index Number:
203-810-4
NACRES:
NA.22
form:
liquid
Assay:
≥98.0% (GC)

vapor pressure

77.8 hPa ( 20 °C)

Quality Level

Assay

≥98.0% (GC)

form

liquid

autoignition temp.

240 °C

potency

>4000 mg/kg LD50, oral (Rat)

expl. lim.

1.1-13.8 % (v/v)

pH

7 (20 °C, 5 g/L in H2O)

bp

85.5 °C/1013 hPa

mp

-70 °C

transition temp

flash point -9 °C

solubility

7.7 g/L

density

0.929 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

O1CCCC=C1

InChI

1S/C5H8O/c1-2-4-6-5-3-1/h2,4H,1,3,5H2

InChI key

BUDQDWGNQVEFAC-UHFFFAOYSA-N

Application

  • 1, 5-Pentanediol production from 3, 4-dihydro-2H-pyran: A novel method for synthesizing bio-based 1,5-pentanediol from 3,4-dihydro-2H-pyran is explored, highlighting its potential industrial application. (Zhao et al., 2024).

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.926 - 0.929
Water (GC): ≤ 0.50 % (a/a)
Identity (IR): passes test

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 3 - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - Skin Sens. 1B

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

5.0 °F - closed cup

Flash Point(C)

-15 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service