Skip to Content
Merck
All Photos(1)

Key Documents

8.01801

Sigma-Aldrich

Benzophenone

for synthesis

Synonym(s):

Benzophenone, Diphenylketon, Diphenylmethanon, Benzoylbenzol

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€28.80
250 G
€39.90
1 KG
€67.60
50 KG
€832.00

€28.80


Estimated to ship on12 May 2025Details


Request a Bulk Order

Select a Size

Change View
5 G
€28.80
250 G
€39.90
1 KG
€67.60
50 KG
€832.00

About This Item

Empirical Formula (Hill Notation):
C13H10O
CAS Number:
Molecular Weight:
182.22
MDL number:
UNSPSC Code:
12352115
EC Index Number:
204-337-6
NACRES:
NA.22

€28.80


Estimated to ship on12 May 2025Details


Request a Bulk Order

vapor pressure

0.004 hPa ( 20 °C)

Quality Level

Assay

≥99.0% (GC)

form

crystals

potency

>10000 mg/kg LD50, oral (Rat)
3535 mg/kg LD50, skin (Rabbit)

reaction suitability

reagent type: reductant

bp

304-306 °C/1013 hPa

mp

47-49 °C

transition temp

flash point 150 °C

density

1.1 g/cm3 at 25 °C

bulk density

700 kg/m3

storage temp.

2-30°C

SMILES string

O=C(c2ccccc2)c1ccccc1

InChI

1S/C13H10O/c14-13(11-7-3-1-4-8-11)12-9-5-2-6-10-12/h1-10H

InChI key

RWCCWEUUXYIKHB-UHFFFAOYSA-N

Application

  • Single and mixture effects of bisphenol A and benzophenone-3: Discusses the immune system effects when exposed to benzophenone-3 and bisphenol A, important for understanding chemical interactions in drug discovery and safety evaluations (Fischer et al., 2024).
  • A surface-independent bioglue using photo-crosslinkable benzophenone moiety: Develops a novel bioglue using benzophenone, which is crucial for applications in medical adhesives and tissue engineering, reflecting the versatility of benzophenone in creating cross-linked polymers (Shi et al., 2024).
  • Photo trapping voltage-gated cardiac sodium channels: Utilizes benzophenone to study local motions within cardiac sodium channels, offering insights into molecular dynamics important for designing drugs targeting cardiovascular diseases (Goodchild and Ahern, 2024).

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Melting range (lower value): ≥ 47 °C
Melting range (upper value): ≤ 49 °C
Identity (IR): passes test
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Carc. 1B - STOT RE 2 Oral

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

280.4 °F - closed cup

Flash Point(C)

138 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service