Skip to Content
Merck
All Photos(1)

Key Documents

8.01391

Sigma-Aldrich

Ethyl vinyl ether

(stabilised with potassium hydroxide) for synthesis

Synonym(s):

Ethyl vinyl ether, Vinyl ethyl ether

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
€35.70
1 L
€81.00
2.5 L
€137.00

€35.70


Estimated to ship on26 March 2025Details


Request a Bulk Order

Select a Size

Change View
100 ML
€35.70
1 L
€81.00
2.5 L
€137.00

About This Item

Linear Formula:
C2H5OCH=CH2
CAS Number:
Molecular Weight:
72.11
MDL number:
UNSPSC Code:
12352112
EC Index Number:
203-718-4
NACRES:
NA.22

€35.70


Estimated to ship on26 March 2025Details


Request a Bulk Order

vapor pressure

560 hPa ( 20 °C)

Quality Level

Assay

≥99.0% (GC)

form

liquid

autoignition temp.

178 °C

potency

6120 mg/kg LD50, oral (Rat)
>15000 mg/kg LD50, skin (Rabbit)

expl. lim.

1.3-12.0 % (v/v)

bp

36 °C/1013 hPa

mp

-115 °C

transition temp

flash point -45 °C

solubility

8.3 g/L

density

0.75 g/cm3 at 20 °C

SMILES string

O(CC)C=C

InChI

1S/C4H8O/c1-3-5-4-2/h3H,1,4H2,2H3

InChI key

FJKIXWOMBXYWOQ-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Density (d 5°C/ 4°C): 0.769 - 0.773
Identity (IR): passes test

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Flam. Liq. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

-49.0 °F

Flash Point(C)

-45 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service