Skip to Content
Merck
All Photos(1)

Key Documents

8.01111

Sigma-Aldrich

4-Aminopyridine

for synthesis

Synonym(s):

4-Aminopyridine, 4-Pyridylamine

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
€85.30

€85.30


Estimated to ship on31 March 2025Details


Request a Bulk Order

Select a Size

Change View
25 G
€85.30

About This Item

Linear Formula:
4-(NH2)C5H4N
CAS Number:
Molecular Weight:
94.11
MDL number:
UNSPSC Code:
12352116
EC Index Number:
207-987-9
NACRES:
NA.22

€85.30


Estimated to ship on31 March 2025Details


Request a Bulk Order

Quality Level

Assay

≥98.0% (GC)

form

crystals

autoignition temp.

640 °C

potency

21 mg/kg LD50, oral (Rat)

bp

274 °C/1013 hPa

mp

157-160 °C

transition temp

flash point 156 °C

solubility

soluble 83 g/L

density

1.26 g/cm3 at 25 °C

storage temp.

2-30°C

SMILES string

Nc1ccncc1

InChI

1S/C5H6N2/c6-5-1-3-7-4-2-5/h1-4H,(H2,6,7)

InChI key

NUKYPUAOHBNCPY-UHFFFAOYSA-N

Application

4-Aminopyridine for synthesis. CAS 504-24-5, chemical formula 4-(NH₂)C₅H₄N.

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Melting range (lower value): ≥ 157 °C
Melting range (upper value): ≤ 160 °C
Identity (IR): passes test

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Oral - Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Aquatic Chronic 2 - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

6.1A - Combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

312.8 °F

Flash Point(C)

156 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service