Skip to Content
Merck
All Photos(1)

Key Documents

8.00821

Sigma-Aldrich

Pentanoic acid

for synthesis

Synonym(s):

Pentanoic acid, Valeric acid

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
€33.80
500 ML
€40.70
1 L
€65.60

€33.80


Estimated to ship on12 May 2025Details


Request a Bulk Order

Select a Size

Change View
100 ML
€33.80
500 ML
€40.70
1 L
€65.60

About This Item

Linear Formula:
CH3(CH2)3COOH
CAS Number:
Molecular Weight:
102.13
MDL number:
UNSPSC Code:
12352106
EC Index Number:
203-677-2
NACRES:
NA.22

€33.80


Estimated to ship on12 May 2025Details


Request a Bulk Order

vapor pressure

0.3 hPa ( 20 °C)

Quality Level

Assay

≥98.0% (GC)

form

liquid

autoignition temp.

360 °C

potency

>2000 mg/kg LD50, skin (Rabbit)

expl. lim.

1.8-7.3 % (v/v)

pH

2.7 (20 °C, 40 g/L in H2O)

bp

186 °C/1013 hPa

mp

-32 °C

transition temp

flash point 86 °C

solubility

40 g/L

density

0.938 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

OC(=O)CCCC

InChI

1S/C5H10O2/c1-2-3-4-5(6)7/h2-4H2,1H3,(H,6,7)

InChI key

NQPDZGIKBAWPEJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.938 - 0.940
Identity (IR): passes test

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible, corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

192.2 °F - closed cup

Flash Point(C)

89 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service