Skip to Content
Merck
All Photos(1)

Key Documents

8.00796

Sigma-Aldrich

Ethanethiol

for synthesis

Synonym(s):

Ethanethiol, Ethyl mercaptan

Sign Into View Organizational & Contract Pricing

Select a Size

250 ML
€45.20
250 ML
€59.90
1 L
€106.00

€45.20


Estimated to ship on22 April 2025Details


Request a Bulk Order

Select a Size

Change View
250 ML
€45.20
250 ML
€59.90
1 L
€106.00

About This Item

Empirical Formula (Hill Notation):
C2H6S
CAS Number:
Molecular Weight:
62.13
MDL number:
UNSPSC Code:
12352105
EC Index Number:
200-837-3
NACRES:
NA.22

€45.20


Estimated to ship on22 April 2025Details


Request a Bulk Order

vapor pressure

576 hPa ( 20 °C)

Quality Level

Assay

≥99% (GC)

form

liquid

autoignition temp.

295 °C

potency

682 mg/kg LD50, oral (Rat)

expl. lim.

2.8-18 % (v/v)

bp

35 °C/1013 hPa

mp

-148 °C

transition temp

flash point -45 °C

density

0.84 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

SCC

InChI

1S/C2H6S/c1-2-3/h3H,2H2,1H3

InChI key

DNJIEGIFACGWOD-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 99 %
Density (d 20 °C/ 4 °C): 0.838 - 0.839
Identity (IR): passes test

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Flam. Liq. 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-49.0 °F - closed cup

Flash Point(C)

-45 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service