Skip to Content
Merck
All Photos(1)

Key Documents

8.00614

Sigma-Aldrich

Pyridine-2,6-dicarboxylic acid

for synthesis

Synonym(s):

Pyridine-2,6-dicarboxylic acid, Dipicolinic acid

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
€52.70
1 KG
€370.00
25 KG
€6,650.00

€52.70


Estimated to ship on07 May 2025Details


Request a Bulk Order

Select a Size

Change View
100 G
€52.70
1 KG
€370.00
25 KG
€6,650.00

About This Item

Linear Formula:
2,6-(HOOC)2C5H3N
CAS Number:
Molecular Weight:
167.12
MDL number:
UNSPSC Code:
12352106
EC Index Number:
207-894-3
NACRES:
NA.22

€52.70


Estimated to ship on07 May 2025Details


Request a Bulk Order

vapor pressure

<0.01 hPa ( 20 °C)

Quality Level

Assay

≥98.0% (acidimetric)

form

powder

autoignition temp.

620 °C

potency

10500 mg/kg LD50, oral (Rat)

pH

2.0 (20 °C, 5 g/L in H2O)

mp

248-255 °C

solubility

5 g/L

bulk density

700 kg/m3

storage temp.

2-30°C

SMILES string

n1c(cccc1C(=O)O)C(=O)O

InChI

1S/C7H5NO4/c9-6(10)4-2-1-3-5(8-4)7(11)12/h1-3H,(H,9,10)(H,11,12)

InChI key

WJJMNDUMQPNECX-UHFFFAOYSA-N

Related Categories

Application


  • Adsorption of Cu(II) ions from aqueous solution using pyridine-2,6-dicarboxylic acid crosslinked chitosan as a green biopolymer adsorbent.: This research explores the efficiency of a green biopolymer, specifically chitosan crosslinked with pyridine-2,6-dicarboxylic acid, for the adsorption of copper ions from aqueous solutions, highlighting its potential for environmental cleanup and water treatment (Bisiriyu et al., 2020).

  • Crystal structure of nonadentate tricompartmental ligand derived from pyridine-2,6-dicarboxylic acid: Spectroscopic, electrochemical and thermal investigations of its transition metal(II) complexes.: This article details the structural and functional characteristics of a complex ligand derived from pyridine-2,6-dicarboxylic acid, which could inform further research in coordination chemistry and material science (Vadavi et al., 2011).

  • Pyridine-substituted oligopeptides as scaffolds for the assembly of multimetallic complexes: variation of chain length.: This research explores the use of pyridine-2,6-dicarboxylic acid in the construction of oligopeptide scaffolds for multimetallic complexes, relevant to the fields of molecular engineering and nanotechnology (Ohr et al., 2005).

  • Comprehensive study of the luminescent properties and lifetimes of Eu(3+) and Tb(3+) chelated with various ligands in aqueous solutions: influence of the synergic agent, the surfactant and the energy level of the ligand triplet.: This study provides a thorough analysis of the luminescent behavior of lanthanide complexes with pyridine-2,6-dicarboxylic acid derivatives, significant for developing advanced photonic and electronic materials (Arnaud et al., 2003).

Analysis Note

Assay (acidimetric): ≥ 98,0 %(m)
Identity (IR): conforms

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible, corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

370.4 °F - closed cup

Flash Point(C)

188 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service