Skip to Content
Merck
All Photos(1)

Key Documents

8.00551

Sigma-Aldrich

Triphenyl phosphite

for synthesis

Synonym(s):

Triphenyl phosphite, Phosphorous acid triphenyl ester

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
€34.90
1 L
€81.70

€34.90


Estimated to ship on31 March 2025Details


Request a Bulk Order

Select a Size

Change View
100 ML
€34.90
1 L
€81.70

About This Item

Linear Formula:
(C6H5O)3P
CAS Number:
Molecular Weight:
310.28
MDL number:
UNSPSC Code:
12352108
EC Index Number:
202-908-4
NACRES:
NA.22

€34.90


Estimated to ship on31 March 2025Details


Request a Bulk Order

vapor pressure

<1 hPa ( 20 °C)

Quality Level

Assay

≥97.0% (GC)

form

solid

pH

1 (20 °C, 200 g/L in H2O)

bp

360 °C/1013 hPa (decomposes)

transition temp

flash point 188 °C

density

1.19 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

P(Oc3ccccc3)(Oc2ccccc2)Oc1ccccc1

InChI

1S/C18H15O3P/c1-4-10-16(11-5-1)19-22(20-17-12-6-2-7-13-17)21-18-14-8-3-9-15-18/h1-15H

InChI key

HVLLSGMXQDNUAL-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 97.0 % (a/a)
Identity (IR): passes test
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 2

Target Organs

Nervous system

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

410.0 °F

Flash Point(C)

210 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service