Skip to Content
Merck
All Photos(1)

Key Documents

8.00500

Sigma-Aldrich

Ethyl butyrate

for synthesis

Synonym(s):

Ethyl butyrate, Butyric acid ethyl ester, Ethyl butanoate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12O2
CAS Number:
Molecular Weight:
116.16
MDL number:
UNSPSC Code:
12352108
EC Index Number:
203-306-4

vapor pressure

17 hPa ( 20 °C)

Quality Level

Assay

≥98.0% (GC)

form

liquid

autoignition temp.

460 °C

potency

13000 mg/kg LD50, oral (Rat)
5228 mg/kg LD50, skin (Rabbit)

bp

121 °C/972 hPa

mp

-98 °C

transition temp

flash point 26 °C

density

0.87 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C6H12O2/c1-3-5-6(7)8-4-2/h3-5H2,1-2H3

InChI key

OBNCKNCVKJNDBV-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.878 - 0.879
Identity (IR): passes test

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

78.8 °F

Flash Point(C)

26 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service