Skip to Content
Merck
All Photos(1)

Key Documents

8.00419

Sigma-Aldrich

2-Aminophenol

for synthesis

Synonym(s):

2-Aminophenol, 2-Hydroxy aniline, 2-Amino-1-hydroxy benzene

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€25.40
250 G
€58.00
1 KG
€103.00
10 KG
€548.00
50 KG
€2,670.00

€25.40


Estimated to ship on05 May 2025Details


Request a Bulk Order

Select a Size

Change View
5 G
€25.40
250 G
€58.00
1 KG
€103.00
10 KG
€548.00
50 KG
€2,670.00

About This Item

Linear Formula:
2-(NH2)C6H4OH
CAS Number:
Molecular Weight:
109.13
MDL number:
UNSPSC Code:
12352104
EC Index Number:
202-431-1
NACRES:
NA.22

€25.40


Estimated to ship on05 May 2025Details


Request a Bulk Order

vapor pressure

14 hPa ( 153 °C)

Quality Level

form

solid

autoignition temp.

190 °C

potency

1300 mg/kg LD50, oral (Rat)

pH

6.5-7.5 (20 °C, 10 g/L in H2O)

bp

150 °C/15 hPa

mp

172-174 °C (sublimed)

transition temp

flash point >175 °C

solubility

17 g/L

density

1.328 g/cm3 at 20 °C

bulk density

600 kg/m3

storage temp.

2-30°C

SMILES string

Nc1c(cccc1)O

InChI

1S/C6H7NO/c7-5-3-1-2-4-6(5)8/h1-4,8H,7H2

InChI key

CDAWCLOXVUBKRW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2-Aminophenol for synthesis. CAS 95-55-6, chemical formula 2-(NH₂)C₆H₄OH.

Analysis Note

Assay (HClO₄): ≥ 99.0 %
Identity (IR): passes test

Pictograms

Exclamation markHealth hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Muta. 2

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

334.4 °F - closed cup

Flash Point(C)

168 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service