Skip to Content
Merck
All Photos(1)

Documents

8.00257

Sigma-Aldrich

3-Chloro-1-propene

(stabilised with propylene oxide) for synthesis

Synonym(s):

3-Chloro-1-propene, Allyl chloride, 3-Chloropropylene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCH2CH=CH2
CAS Number:
Molecular Weight:
76.52
MDL number:
UNSPSC Code:
12352101
EC Index Number:
203-457-6
NACRES:
NA.22

vapor pressure

400 hPa ( 20 °C)

Quality Level

Assay

≥98.0% (GC)

form

liquid

autoignition temp.

390 °C

potency

450 mg/kg LD50, oral (Rat)
540 mg/kg LD50, skin (Rat)

expl. lim.

3.3-11.2 % (v/v)

mp

-134.5 °C (ECHA)

transition temp

flash point -32 °C

solubility

3.6 g/L

density

0.94 g/cm3 at 20 °C

storage temp.

2-8°C

InChI

1S/C3H5Cl/c1-2-3-4/h2H,1,3H2

InChI key

OSDWBNJEKMUWAV-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.936 - 0.940
Identity (IR): passes test
The material is stabilised with propylene oxide. At time of analytical testing a content of >300 ppm stabiliser was determined.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Carc. 2 - Eye Irrit. 2 - Flam. Liq. 2 - Muta. 2 - Skin Irrit. 2 - STOT RE 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

-25.6 °F

Flash Point(C)

-32 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service