Skip to Content
Merck
All Photos(1)

Key Documents

71789

Sigma-Aldrich

Overnight Express Autoinduction NMR Medium - 15N, 13C - Novagen

Sign Into View Organizational & Contract Pricing

Select a Size

71789-3
€556.00

€556.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
71789-3
€556.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.54

€556.00


Please contact Customer Service for Availability

Request a Bulk Order

manufacturer/tradename

Novagen®

Quality Level

storage condition

OK to freeze

General description

Overnight Express™ Autoinduction NMR-15N, -13C, when supplemented with 13C-labeled glycerol (Cat. No. CLM1510EMD), provides reagents to make 1 L of defined autoinducing liquid culture medium used to produce [U-15N, U-13C]-labeled proteins for NMR spectroscopy. The autoinduction method is based on the differential metabolism of carbon sources to promote growth to high cell density and to automatically induce protein expression under control of lac promoters, eliminating the need to monitor cell densities or add inducer (IPTG). The system includes four concentrated sterile solutions: OnEx Dual-NMR Solution 1 is a blend of carbon sources optimized for tightly regulated uninduced growth to high density, followed by induction; OnEx NMR Solution 2 is a buffer and nitrogen blend that mediates metabolic acid production and provides additional nitrogen for increased protein synthesis; OnEx Solution 3 provides magnesium for maximal cell density; OnEx Solution 4 provides trace metals to minimize growth limitations associated with mineral deficiencies and satisfy the requirements of metal-containing target proteins, even at high expression levels. The kit also includes 15N-labeled solid ammonium chloride, 13C-labeled solid glucose, and reagents to make a non-inducing starter medium, for staged growth without expression and for stable storage of clones. Efficient 13C protein labeling requires 13C-glycerol, which must be purchased separately. Addition of the four liquid components and sterile solutions of 15N ammonium chloride, 13C glucose, and 13C glycerol to sterile water results in a chemically- defined medium promoting high cell densities, autoinduction of expression, and efficient labeling of target proteins.

Warning

Toxicity: Harmful

Storage and Stability

Ship Code: Multiple Storage Conditions
Storage: Multiple storage requirements

Legal Information

Express is a trademark of Merck KGaA, Darmstadt, Germany
NOVAGEN is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 1B - Eye Irrit. 2 - Met. Corr. 1 - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service