Skip to Content
Merck
All Photos(1)

Key Documents

71554

Sigma-Aldrich

pET-52b(+) DNA - Novagen

Novagen′s pET-52b(+) vector is designed for cloning and high-level expression of proteins fused with the 8 aa Strep-Tag II coding sequence that is cleavable with human rhinovirus (HRV) 3C protease.

Synonym(s):

Novagen′s pET-52b(+) DNA Vector, pET Vectors, pET- 52b(+) vector

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
€328.00

€328.00


In StockDetails


Request a Bulk Order

Select a Size

Change View
10 μG
€328.00

About This Item

UNSPSC Code:
41106610

€328.00


In StockDetails


Request a Bulk Order

Quality Level

manufacturer/tradename

Novagen®

storage condition

OK to freeze

shipped in

dry ice
wet ice

storage temp.

−70°C

General description

The pET-52b(+) vector is designed for cloning and high-level expression of target proteins fused with the 8 aa Strep•Tag II coding sequence that is cleavable with human rhinovirus (HRV) 3C protease. The plasmid contains a strong T7lac promoter, an optimized RBS, the coding sequence for the HRV 3C protease cleavage site (LysGluValLysPheGln↓GlyPro), and a multiple cloning site that contains restriction enzyme sites found in many other Novagen expression vectors to facilitate insert transfer. It also encodes an optional C-terminal thrombin protease cleavage site and HIS TAG® coding sequence with 10 histidines. The HIS TAG fusion is compatible with purification, detection, and quantification.

The pET Vectors are supplied as purified plasmid DNA (10 µg). Each order of pET DNA also includes an Induction Control strain (supplied as a glycerol stock). Please contact technical service if you need additional information.

Warning

Toxicity: Standard Handling

Legal Information

HIS TAG is a registered trademark of Merck KGaA, Darmstadt, Germany
NOVAGEN is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

This product is sold for internal research use only. Any commercial use of this product, its components, and/or any derivatives thereof (including but not limited to proteins produced using the product or its components) (together and hereinafter the ′EMD Product′) requires signature of a written commercial use agreement with EMD Millipore Corporation or its successor-in-interest. Commercial use shall include but not be limited to: (1) use of the EMD Product to manufacture products for sale to third parties; (2) use of the EMD Product to provide services, information, or data to third parties in exchange for consideration; (3) use of the EMD Product for therapeutic, diagnostic or prophylactic purposes (including as part of a device, chip, assay or other product); or (4) resale of the EMD Product, whether or not such EMD Product is resold for research use. Nothing contained herein shall be deemed to represent or warrant that additional third party rights are not required for use of the EMD Product. Please direct any questions on these use restrictions to: [email protected].

This product contains genetically modified organisms (GMO). Within the EU GMOs are regulated by Directives 2001/18/EC and 2009/41/EC of the European Parliament and of the Council and their national implementation in the member States respectively. This legislation obliges MilliporeSigma to request certain information about you and the establishment where the GMOs are being handled. Clickhere for Enduser Declaration (EUD) Form.

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service