Skip to Content
Merck
All Photos(1)

Key Documents

71545

Sigma-Aldrich

BacMagic DNA Kit - Novagen

Synonym(s):

BacMagic-1 DNA Kit

Sign Into View Organizational & Contract Pricing

Select a Size

5 REACTIONS
€794.00

€794.00


Estimated to ship on22 April 2025Details


Request a Bulk Order

Select a Size

Change View
5 REACTIONS
€794.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.51

€794.00


Estimated to ship on22 April 2025Details


Request a Bulk Order

manufacturer/tradename

Novagen®

Quality Level

storage condition

OK to freeze

shipped in

wet ice

storage temp.

2-8°C

General description

The BacMagic-1 DNA Kit provides a positive selection for creating baculovirus recombinants in preparation for generating recombinant protein expression in insect cells. The kit improves on the traditional method for generating recombinant baculoviruses by eliminating the tedious, time-consuming steps of plaque purification. BacMagic DNA is an AcNPV genome with a portion of the essential open reading frame (ORF) 1629 deleted, preventing nonrecombinant virus from replicating in insect cells. The target coding sequence in a compatible transfer plasmid, such as pIEx/Bac, pBAC, or pTriEx vectors, is cotransfected with BacMagic DNA into insect cells. Homologous recombination within the cells restores the function of the viral ORF1629. Only recombinant baculovirus can replicate, producing a homogeneous population of recombinants. The BacMagic™-1 DNA Kit contains the BacMagic-1 DNA, Insect GeneJuice® Transfection Reagent, and a positive control plasmid to monitor co-transfection efficiency.

Components

  • 0.5 µg- BacMagic DNA
  • 50 µl- Insect GeneJuice Transfection Reagent
  • 2 µg- Transfection Control Plasmid

Warning

Toxicity: Multiple Toxicity Values, refer to MSDS

Legal Information

NOVAGEN is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service