Skip to Content
Merck
All Photos(1)

Key Documents

71511

Millipore

D-Tube Electroelution Accessory Kit

Sign Into View Organizational & Contract Pricing

Select a Size

1 KIT
€105.00

€105.00


In StockDetails



Select a Size

Change View
1 KIT
€105.00

About This Item

UNSPSC Code:
41104900
NACRES:
NB.24

€105.00


In StockDetails


manufacturer/tradename

Novagen®

Quality Level

storage condition

OK to freeze

shipped in

ambient

storage temp.

10-30°C

Components

•: 1: Supporting Tray, Mini


•: 1: Supporting Tray, Midi


•: 1: Supporting Tray, Maxi


•: 1 ml: MS Precipitation Buffer


•: 10 ml: Tricholoroacetic acid, 20% w/v


•: 2 x 1 ml: 3M Sodium acetate, pH 5.2

Warning

Toxicity: Multiple Toxicity Values, refer to MSDS (O)

Other Notes

Due to the nature of the Hazardous Materials in this shipment, additional shipping charges may be applied to your order. Certain sizes may be exempt from the additional hazardous materials shipping charges. Please contact your local sales office for more information regarding these charges.

Legal Information

NOVAGEN is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Dam. 1 - Skin Corr. 1A - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service