Skip to Content
Merck
All Photos(1)

Key Documents

71074

Millipore

BetaBlue Staining Kit

Convenient visualization of β-gal in cells or tissues

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
€267.00

€267.00


Please contact Customer Service for Availability


Select a Size

Change View
100 ML
€267.00

About This Item

UNSPSC Code:
41106513
NACRES:
NA.31

€267.00


Please contact Customer Service for Availability

manufacturer/tradename

Novagen®

storage condition

OK to freeze

shipped in

wet ice

General description



Convenient visualization of β-gal in cells or tissues
The BetaBlue Staining Kit provides direct visualization of β-galactosidase reporter expression in isolated cells, tissues, or intact organisms. The kit contains sufficient reagents for 100 staining reactions and includes solutions of the substrate X-Gal (5-bromo-4-chloro-3-indolyl-β-D-galactopyranoside) and Reaction Buffer optimized for rapid, sensitive histochemical staining with minimal background. The exceptional staining seen with the BetaBlue Staining Kit enables quick, accurate determination of transfection efficiencies, assessment of stable cell line generation, and transgene expression in tissue slices or whole mounts of transgenic animals.

Components

•2 × 50 mlBetaBlue Reaction Buffer

•3 × 1 mlBetaBlue X-gal Solution

Warning

Toxicity: Multiple Toxicity Values, refer to MSDS (O)

Legal Information

NOVAGEN is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sebastian Frische et al.
American journal of physiology. Renal physiology, 320(1), F74-F86 (2020-12-08)
Variations in the claudin-14 (CLDN14) gene have been linked to increased risk of hypercalciuria and kidney stone formation. However, the exact cellular localization of CLDN14 and its regulation remain to be fully delineated. To this end, we generated a novel
Keishi Otsu et al.
Journal of bone and mineral research : the official journal of the American Society for Bone and Mineral Research, 31(11), 1943-1954 (2016-10-27)
During tooth development, oral epithelial cells differentiate into ameloblasts in order to form the most mineralized tissue in the vertebrate body: enamel. During this process, ameloblasts directionally secrete enamel matrix proteins and morphologically change from low columnar cells to polarized

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service