Skip to Content
Merck
All Photos(1)

Key Documents

70979

Sigma-Aldrich

BetaFluor β-Galactosidase Assay Kit

Optimized reagents for ultrasensitive β-gal assays

Sign Into View Organizational & Contract Pricing

Select a Size

500 EA
€851.00

€851.00


Estimated to ship on17 February 2025Details



Select a Size

Change View
500 EA
€851.00

About This Item

UNSPSC Code:
41106513
NACRES:
NA.54

€851.00


Estimated to ship on17 February 2025Details


Quality Level

manufacturer/tradename

Novagen®

storage condition

OK to freeze

storage temp.

−20°C

General description

Optimized reagents for ultrasensitive β-gal assays
The BetaFluor β-Gal Assay Kit is designed for rapid, sensitive measurement of β-galactosidase activity in cell extracts. The kit features Reportasol Extraction Buffer, which enables efficient passive extraction of cell protein while maintaining optimal enzyme activity, and is configured for 96-well format. Because no mechanical operations are necessary for extraction, the assay is highly amenable to high-throughput reporter gene expression analysis. The BetaFluor Assay is designed for fluorometric detection with 500-fg sensitivity.

Components

•100 mgBetaFluor Substrate

•25 mlReportasol Extraction Buffer

•100 mlReaction Buffer

•50 mlStop Buffer

•1.6 ml1 M DTT

Warning

Toxicity: Multiple Toxicity Values, refer to MSDS (O)

Legal Information

NOVAGEN is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Justine M Pinskey et al.
The Journal of biological chemistry, 292(37), 15192-15204 (2017-07-02)
Hedgehog (HH) signaling critically regulates embryonic and postnatal development as well as adult tissue homeostasis, and its perturbation can lead to developmental disorders, birth defects, and cancers. Neuropilins (NRPs), which have well-defined roles in Semaphorin and VEGF signaling, positively regulate
Martin F Engelke et al.
Current biology : CB, 29(7), 1137-1148 (2019-03-25)
The trafficking of components within cilia, called intraflagellar transport (IFT), is powered by kinesin-2 and dynein-2 motors. Loss of function in any subunit of the heterotrimeric KIF3A/KIF3B/KAP kinesin-2 motor prevents ciliogenesis in mammalian cells and has hindered an understanding of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service