Skip to Content
Merck
All Photos(1)

Key Documents

70608

Sigma-Aldrich

pETBlue-1 DNA - Novagen

Novagen′s pETBlue-1 vector is designed to identify recombinants by traditional blue/white screening while also allowing T7lac promoter based expression of target genes.

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
€418.00

€418.00


Estimated to ship on02 May 2025Details



Select a Size

Change View
20 μG
€418.00

About This Item

UNSPSC Code:
41106610
NACRES:
NA.51

€418.00


Estimated to ship on02 May 2025Details


Quality Level

form

liquid

manufacturer/tradename

Novagen®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

shipped in

wet ice

storage temp.

−20°C

General description

The pETBlue-1 vector is designed to identify recombinants by traditional blue/white screening while also allowing T7lac promoter based expression of target genes. Screening is independent of expression because the T7lac expression promoter is in an opposed orientation relative to the E. coli promoter that mediates blue/white screening. pETBlue-1 facilitates the expression of native unfused proteins and allows convenient subcloning of target genes already fused to existing detection and purification tags. The EcoR V cloning site is appropriately spaced downstream of an E. coli ribosome binding site. Inserts must encode an ATG start codon at their 5′ end if expression is desired. The sequence is numbered from the first base of the T7 promoter sequence.

Warning

Toxicity: Standard Handling (A)

Legal Information

NOVAGEN is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service