Skip to Content
Merck
All Photos(1)

Key Documents

693019

Millipore

Zwittergent® 3-08 Detergent

A synthetic zwitterionic detergent, which unlike other amphoteric surfactants, retains its zwitterionic character over a broad pH range.

Synonym(s):

n-Octyl-N,N-dimethyl-3-ammonio-1-propanesulfonate, hplc solvents

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€186.00

€186.00


Estimated to ship on05 May 2025Details


Request a Bulk Order

Select a Size

Change View
5 G
€186.00

About This Item

Empirical Formula (Hill Notation):
C13H29NO3S
CAS Number:
Molecular Weight:
279.44
MDL number:
UNSPSC Code:
12161902
NACRES:
NA.21

€186.00


Estimated to ship on05 May 2025Details


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated

color

white

CMC

330 mM

solubility

water: soluble

shipped in

ambient

storage temp.

15-25°C

SMILES string

[S](=O)(=O)([O-])CCC[N+](CCCCCCCC)(C)C

InChI

1S/C13H29NO3S/c1-4-5-6-7-8-9-11-14(2,3)12-10-13-18(15,16)17/h4-13H2,1-3H3

InChI key

QZRAABPTWGFNIU-UHFFFAOYSA-N

General description

A synthetic zwitterionic detergent, which unlike other amphoteric surfactants, retains its zwitterionic character over a broad pH range (CMC: 330 mM). This property is attributed to both the presence of a strongly basic quaternary ammonium ion and an acidic sulfonate ion of equal strength. This ionic balance presumably prevents irreversible binding to either anionic or cationic compounds.

Application


  • Use of a zwitterionic detergent for the restoration of the antibody binding capacity of immunoblotted Francisella tularensis lipopolysaccharide.: This study explores the application of Zwittergent 3-08 detergent in the restoration of antibody binding capacity to lipopolysaccharides of Francisella tularensis, enhancing the immunoblotting technique. This finding is significant for biochemical assays involving bacterial lipopolysaccharides (Fulop et al., 1992, DOI).

  • The effect of zwitterionic detergents on the extraction and functional properties of cartilage proteoglycans.: This research demonstrates how Zwittergent 3-08 detergent affects the extraction and functionality of cartilage proteoglycans, providing insights into its potential uses in cartilage biochemistry and proteoglycan studies (Byers et al., 1987, PubMed).

  • Characterization of covalently cross-linked pancreatic somatostatin receptors.: This paper describes the use of Zwittergent 3-08 detergent in characterizing pancreatic somatostatin receptors, highlighting its role in receptor biochemistry and cross-linking studies (Susini et al., 1986, PubMed).

Packaging

Packaged under inert gas

Warning

Toxicity: Irritant (B)

Reconstitution

Following reconstitution, aliquot and refrigerate (4°C). Stock solutions are stable for up to 1 month at 4°C.

Other Notes

Fulop, M.J., et al. 1992. Anal. Biochem. 203, 141.
Byers, S., et al. 1987. J. Biol. Chem.262, 9166.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
Zwittergent is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service