Skip to Content
Merck
All Photos(1)

Key Documents

682151

Sigma-Aldrich

Xanthine Oxidase, Buttermilk

Native xanthine oxidase from buttermilk. Complex metalloprotein that catalyzes oxidative hydroxylation of a variety of aromatic heterocycles and simple aldehydes. Catalyzes the oxidation of xanthine to uric acid.

Synonym(s):

Xanthine Oxidase, Buttermilk

Sign Into View Organizational & Contract Pricing

Select a Size

10 U
€113.00
50 U
€316.00

€113.00


Estimated to ship on21 May 2025Details



Select a Size

Change View
10 U
€113.00
50 U
€316.00

About This Item

CAS Number:
Enzyme Commission number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.54

€113.00


Estimated to ship on21 May 2025Details


Quality Level

description

RTECS - RQ8455000

form

liquid

specific activity

≥0.5 units/mg protein

manufacturer/tradename

Calbiochem®

storage condition

do not freeze

foreign activity

lactoperoxidase, none detected

shipped in

wet ice

storage temp.

2-8°C

General description

Native xanthine oxidase from buttermilk. Complex metalloprotein that catalyzes oxidative hydroxylation of a variety of aromatic heterocycles and simple aldehydes. Catalyzes the oxidation of xanthine to uric acid.

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Standard Handling (A)

Unit Definition

One unit is defined as the amount of enzyme that will convert 1.0 µmol of xanthine to uric acid per min at 25°C, pH 7.5.

Physical form

Crystalline suspension in 60% ammonium sulfate.

Other Notes

McWhirter, R.B. and Hille, R. 1991. J. Biol. Chem.266, 23724.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Synthesis of Melting Gels Using Mono-Substituted and Di-Substituted Alkoxysiloxanes

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service