Skip to Content
Merck
All Photos(1)

Key Documents

681640

Sigma-Aldrich

Wee1 Inhibitor

The Wee1 Inhibitor, also referenced under CAS 622855-37-2, controls the biological activity of Wee1. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

Wee1 Inhibitor, 4-(2-Chlorophenyl)-9-hydroxypyrrolo[3,4-c]carbazole-1,3-(2H,6H)-dione, Chk1 Inhibitor V

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€176.00

€176.00


Estimated to ship on24 March 2026Details



Select a Size

Change View
1 MG
€176.00

About This Item

Empirical Formula (Hill Notation):
C20H11ClN2O3
CAS Number:
Molecular Weight:
362.77
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€176.00


Estimated to ship on24 March 2026Details


Quality Level

Assay

≥97% (HPLC)

form

solid

potency

11 nM IC50

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

light orange

solubility

DMSO: 10 mg/mL
ethanol: 10 mg/mL

shipped in

ambient

storage temp.

−20°C

SMILES string

Clc1c(cccc1)c2c3c(c4c([nH]c5c4cc(cc5)O)c2)[c]([nH][c]3=O)=O

InChI

1S/C20H11ClN2O3/c21-13-4-2-1-3-10(13)11-8-15-16(18-17(11)19(25)23-20(18)26)12-7-9(24)5-6-14(12)22-15/h1-8,22,24H,(H,23,25,26)

InChI key

DPEXRCOBPACFOO-UHFFFAOYSA-N

General description

A pyrrolocarbazole compound that acts as a potent, ATP-binding site-targeting inhibitor of Wee1 (IC50 = 11 nM) in an ATP-competitive manner and displays ~40-fold selectivity over the related checkpoint kinase Chk1 (IC50 = 440 nM).

Biochem/physiol Actions

Cell permeable: no
Product competes with ATP.
Reversible: no

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service