Skip to Content
Merck
All Photos(1)

Key Documents

676377

Sigma-Aldrich

Valinomycin, Streptomyces fulvissimus

A cyclododecadepsi-peptide ionophore antibiotic.

Synonym(s):

Valinomycin, Streptomyces fulvissimus

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C54H90N6O18
CAS Number:
Molecular Weight:
1111.32
MDL number:
UNSPSC Code:
12352200

Quality Level

Assay

≥93% (HPLC)

form

crystalline solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

solubility

DMSO: soluble
acetic acid: soluble
chloroform: soluble
ether: soluble

shipped in

ambient

storage temp.

10-30°C

InChI

1S/C54H90N6O18/c1-22(2)34-49(67)73-31(19)43(61)55-38(26(9)10)53(71)77-41(29(15)16)47(65)59-36(24(5)6)51(69)75-33(21)45(63)57-39(27(11)12)54(72)78-42(30(17)18)48(66)60-35(23(3)4)50(68)74-32(20)44(62)56-37(25(7)8)52(70)76-40(28(13)14)46(64)58-34/h22-42H,1-21H3,(H,55,61)(H,56,62)(H,57,63)(H,58,64)(H,59,65)(H,60,66)/t31-,32-,33-,34+,35+,36?,37-,38-,39-,40+,41+,42+/m0/s1

InChI key

FCFNRCROJUBPLU-UYBNATROSA-N

General description

A cyclododecadepsi-peptide ionophore antibiotic. Potassium ionophore that transports alkali metal ions across artificial or biological lipid membranes (Rb+ >K+ >Cs+ >Ag+ >NH4+ >Na+ >Li+). Induces K+ conductivity in cell membranes at concentrations as low as 10 nM. Because of its ability to affect membrane transport of K+ and to create a hydrophobic layer on solid electrodes, it is often used in membrane electrode systems for determining K+ concentration. Useful in studies of K+ transport in mitochondria. Uncouples oxidative phosphorylation by binding to sites on membranes rich in sulfhydryl groups. Induces apoptosis in murine thymocytes. Also reported to inhibit NGF-induced neuronal differentiation.

Biochem/physiol Actions

Cell permeable: no
Primary Target
NGF-induced neuronal differentiation
Product does not compete with ATP.
Reversible: no

Warning

Toxicity: Highly Toxic (H)

Other Notes

Due to the nature of the Hazardous Materials in this shipment, additional shipping charges may be applied to your order. Certain sizes may be exempt from the additional hazardous materials shipping charges. Please contact your local sales office for more information regarding these charges.
Harada, H., et al. 1994. Biochim. Biophys. Acta1220, 310.
Luvisetto, S., et al. 1994. Biochim. Biophys. Acta 1186, 12.
Orlov, V.N., et al. 1994. FEBS Lett.345, 104.
Acra, S., et al. 1993. Am. J. Physiol.264, G45.
Cornet, M., et al. 1993. J. Membr. Biol.131, 55.
Deckers, C.L., et al. 1993. Exp. Cell Res.208, 362.
Dimroth, P., and Thomer, A. 1993. Biochemistry32, 1734.
Opanasenko, V.K., et al. 1992. FEBS Lett.307, 280.
Pape, P.C., et al. 1992. J. Physiol.449, 219.
Ahmed, S., and Booth, I.R. 1983. Biochem. J.212, 105.
Lerner, A., et al. 1982. FEBS Lett.146, 9.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 1 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service