Skip to Content
Merck
All Photos(1)

Key Documents

666133

Sigma-Aldrich

Urease, Canavalia ensiformis

Native urease from Canavalia ensiformis. Catalyzes hydrolysis of urea to carbon dioxide and ammonia. Useful in the determination of urea in body fluids. Note: 1 KU = 1000 units.

Synonym(s):

Urease, Canavalia ensiformis

Sign Into View Organizational & Contract Pricing

Select a Size

10 KU
€94.30

€94.30


Estimated to ship on23 April 2025Details



Select a Size

Change View
10 KU
€94.30

About This Item

CAS Number:
Enzyme Commission number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.54

€94.30


Estimated to ship on23 April 2025Details


biological source

Canavalia ensiformis

Quality Level

description

Merck USA index - 14, 9870

form

lyophilized solid

specific activity

>150 units/mg material (Nesslers)

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

solubility

0.05 M Tris-HCl/0.001 M EDTA, pH 8.0: 5 mg/mL

cation traces

NH4+: ≤4 μmol/U (μmol/U enzyme)

shipped in

ambient

storage temp.

−20°C

General description

Native urease from Canavalia ensiformis. Catalyzes hydrolysis of urea to carbon dioxide and ammonia. Useful in the determination of urea in body fluids. Note: 1 KU = 1000 units.

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Standard Handling (A)

Unit Definition

One unit is defined as the amount of enzyme that will release 1 µmol of ammonia from urea per min at 25°C, pH 7.0.

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 2 months at -20°C.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service