Skip to Content
Merck
All Photos(1)

Key Documents

662006

Sigma-Aldrich

U0124

A useful negative control for MEK inhibitors U0125 and U0126.

Synonym(s):

U0124, 1,4-Diamino-2,3-dicyano-1,4- bis(methylthio)butadiene, 1,4-Diamino-2,3-dicyano-1,4-bis(methylthio)butadiene

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€123.00

€123.00


Estimated to ship on08 April 2025Details



Select a Size

Change View
1 MG
€123.00

About This Item

Empirical Formula (Hill Notation):
C8H10N4S2
CAS Number:
Molecular Weight:
226.32
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€123.00


Estimated to ship on08 April 2025Details


Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 100 mg/mL

shipped in

ambient

storage temp.

−20°C

General description

A useful negative control for MEK inhibitors U0125 (Cat. No. 662008) and U0126 (Cat. No. 662005). Does not inhibit MEK activity even at concentrations of 100 µM.
A useful negative control for the MEK inhibitors U0125 (Cat. No. 662008) and U0126 (Cat. No. 662005). Does not inhibit MEK even at concentrations of 100 µM.

Biochem/physiol Actions

Cell permeable: no
Primary Target
MEK1
Product does not compete with ATP.
Reversible: no

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 2 weeks at -20°C.

Other Notes

Favata, M.F., et al. 1998. J. Biol. Chem. 273, 18623.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Katherine E Savell et al.
Science advances, 6(26), eaba4221-eaba4221 (2020-07-09)
Drugs of abuse elevate dopamine levels in the nucleus accumbens (NAc) and alter transcriptional programs believed to promote long-lasting synaptic and behavioral adaptations. Here, we leveraged single-nucleus RNA-sequencing to generate a comprehensive molecular atlas of cell subtypes in the NAc

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service