Skip to Content
Merck
All Photos(2)

Key Documents

648451

Sigma-Aldrich

Trk Inhibitor III, GNF-5837

The Trk Inhibitor III, GNF-5837 controls the biological activity of Trk. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

Trk Inhibitor III, GNF-5837, (Z)-1-(3-((3-((1H-Pyrrol-2-yl)methylene)-2-oxoindolin-6-yl)amino)-4-methylphenyl)-3-(2-fluoro-5-(trifluoromethyl)phenyl)urea

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€294.00

€294.00


Estimated to ship on07 April 2026Details



Select a Size

Change View
10 MG
€294.00

About This Item

Empirical Formula (Hill Notation):
C28H21F4N5O2
CAS Number:
Molecular Weight:
535.49
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€294.00


Estimated to ship on07 April 2026Details


Quality Level

Assay

≥98% (HPLC)

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

orange

solubility

DMSO: 50 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

O=C(NC1=C(C=CC(C(F)(F)F)=C1)F)NC2=CC(NC3=CC(NC/4=O)=C(C=C3)C4=C\C5=CC=CN5)=C(C)C=C2

InChI

1S/C28H21F4N5O2/c1-15-4-6-19(35-27(39)37-25-11-16(28(30,31)32)5-9-22(25)29)13-23(15)34-18-7-8-20-21(12-17-3-2-10-33-17)26(38)36-24(20)14-18/h2-14,33-34H,1H3,(H,36,38)(H2,35,37,39)/b21-12-

InChI key

YYDUWLSETXNJJT-MTJSOVHGSA-N

General description

An orally bioavailable oxindole compound that and acts as a potent, reversible and type II DFG-out inhibitor of pan-Trk activity (IC50 = 8 and 12 nM for TrkA and TrkB). Shown to target Trk (tropomyosin receptor kinase) ATP binding cleft and an immediately adjacent hydrophobic pocket. Preferentially arrests the proliferation of Ba/F3 cells fused with Tel-TrkA, Tel-TrkB and Tel-TrkC (IC50 = 11, 9 and 7 nM, respectively) and in Ba/F3 and RIE cells expressing both TrkA and NGF (IC50 = 42 and 17 nM, respectively) over Mo7e-c-Kit and Rat-A10-PDGFR (IC50 = 1 and 0.5 µM) and Ba/F3-Tel-KDR and wt-Ba/F3 cells (IC50 = 3.0 and 5.6 µM). Displays ~100-fold greater selectivity among a panel of 59 closely related kinases and in 33 cellular kinase assays. Weakly active against relevant cytochrome P450 isozymes and hERG channel, and exhibit adequate microsomal stability, pharmacokinetic profile and efficacy in mice and rats. Suppresses tumor growth in a mouse RIE-TrkAmNGF xenograft model (50 mg/kg, p.o.).

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Albaugh, P., et al. 2012. ACS Med. Chem. Lett.3, In press.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service