Skip to Content
Merck
All Photos(2)

Key Documents

616521

Sigma-Aldrich

TRPV4 Antagonist II, HC-067047

The TRPV4 Antagonist II, HC-067047 controls the biological activity of TRPV4.

Synonym(s):

TRPV4 Antagonist II, HC-067047, 2-Methyl-1-(3-morpholinopropyl)-5-phenyl-N-(3-(trifluoromethyl)phenyl)-1H-pyrrole-3-carboxamide, Transient Receptor Potential Vanilloid-4 Antagonist II, HC067047, HC067047, 2-Methyl-1-(3-morpholinopropyl)-5-phenyl-N-(3-(trifluoromethyl)phenyl)-1H-pyrrole-3-carboxamide, Transient Receptor Potential Vanilloid-4 Antagonist II

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€171.00

€171.00


Estimated to ship on05 May 2025Details


Request a Bulk Order

Select a Size

Change View
10 MG
€171.00

About This Item

Empirical Formula (Hill Notation):
C26H28F3N3O2
CAS Number:
Molecular Weight:
471.51
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€171.00


Estimated to ship on05 May 2025Details


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 50 mg/mL, clear, colorless

shipped in

ambient

storage temp.

2-8°C

SMILES string

FC(F)(F)c1cc(ccc1)NC(=O)c2c([n](c(c2)c4ccccc4)CCCN3CCOCC3)C

InChI key

NCZYSQOTAYFTNM-UHFFFAOYSA-N

General description

A pyrrolocarboxamide compound that potently antagonizes TRP (transient receptor potential) family cation channel TRPV4 activity (IC50 = 17, 48, and 133 nM, respectively, against mouse, human, or rat TRPV4 in whole cell voltage-gated current measurement) in a reversible manner, displaying much reduced potency against human ERG and TRPM8 (IC50 = 368 and 780 nM, respectively) and little or no activity toward hKv1.3, hTRPA1, hTRPC5, hTRPV1/2/3/6, mTRPM7, mTRPV2, or rKv1.2. HC-067047 is efficacious (10 mg/kg via i.p.) in reducing micturition (urination) frequency and increasing voided urine volume of mice and rats suffering from CPA- (Cat. No. 239785) induced cystitis in vivo.

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Everaerts, W., et al. 2010. Proc. Natl. Acad. Sci. USA107, 19084.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service