Skip to Content
Merck
All Photos(1)

Documents

616430

Sigma-Aldrich

TGF-α, Human, Recombinant, E. coli

Synonym(s):

TGF-α, Human, Recombinant, E. coli, Transforming Growth Factor-α

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352202

Assay

≥97% (SDS-PAGE)

Quality Level

form

lyophilized

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

impurities

≤100 pg/μg Endotoxin (pg/μg TGF-α)

shipped in

dry ice

storage temp.

−70°C

General description

Recombinant, human transforming growth factor-α expressed in E. coli. Plays an important role in cell-cell adhesion and cell growth and differentiation. Reported to play a role in the development of hormonal insensitivity in estrogen-receptor-positive breast cancers.

Biochem/physiol Actions

ED₅₀= 0.1-0.4 ng/ml as measured by the ability to stimulate ³H-thymidine incorporation in the mouse fibroblast cell line Balb/3T3

Warning

Toxicity: Standard Handling (A)

Physical form

Lyophilized from 30% acetonitrile, 0.1% TFA.

Reconstitution

Following reconstitution, aliquot and freeze (-70°C) for long-term storage or refrigerate (4°C) for short-term storage. Avoid freeze/thaw cycles of solutions. Stock solutions are stable for up to 1 month at 4°C or for up to 3 months at -70°C.
Reconstitute to a concentration of ≥10 µg/ml with sterile 10 mM acetic acid containing ≥0.1% HSA or BSA.

Other Notes

Nicholson, R.I., et al. 1994. Cancer Res. 54, 1684.
Anklesaria, P., et al. 1990. Proc. Natl. Acad. Sci. USA87, 3289.
Massague, J. 1990. J. Biol. Chem.265, 21393.
Watanabe, S., et al. 1987. Proc. Natl. Acad. Sci. USA84, 1258.
Schreiber, A.B., et al. 1986. Science232, 1250.
Tam, J.P. 1985. Science229, 673.
Derynck, R., et al. 1984. Cell38, 327.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service