Skip to Content
Merck
All Photos(1)

Key Documents

616394

Millipore

TPEN

Synonym(s):

TPEN, N,N,Nʹ,Nʹ- tetrakis-(2-Pyridylmethyl)ethylenediamine

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
€181.00

€181.00


Estimated to ship on05 May 2025Details


Request a Bulk Order

Select a Size

Change View
100 MG
€181.00

About This Item

Empirical Formula (Hill Notation):
C26H28N6
CAS Number:
Molecular Weight:
424.54
MDL number:
UNSPSC Code:
12352116
NACRES:
NA.25

€181.00


Estimated to ship on05 May 2025Details


Request a Bulk Order

Assay

≥97% (titration)

Quality Level

form

solid

reaction suitability

reagent type: chelator

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

tan

solubility

ethanol: 10 mg/mL
0.1 M phosphate pH 7.0: 25 μM
DMSO: soluble

shipped in

ambient

storage temp.

15-25°C

SMILES string

N(CCN(Cc4ncccc4)Cc3ncccc3)(Cc2ncccc2)Cc1ncccc1

InChI

1S/C26H28N6/c1-5-13-27-23(9-1)19-31(20-24-10-2-6-14-28-24)17-18-32(21-25-11-3-7-15-29-25)22-26-12-4-8-16-30-26/h1-16H,17-22H2

InChI key

CVRXLMUYFMERMJ-UHFFFAOYSA-N

General description

Lipid-soluble heavy metal chelator that crosses artificial and natural membranes. Auxiliary reagent in certain techniques used for measuring free Ca2+ levels in cells. Also useful in extracting Aβ amyloid deposits from Alzheimer’s disease brain tissue. Has extremely high affinity for heavy metals but low affinity for Ca2+ and Mg2+. Induces DNA fragmentation when chelated with Zn2+ and induces apoptosis in thymocytes. Strips heavy metals from liposome-trapped QUIN 2 (Cat. No. 551826).

Application


  • Poly(ADP-ribosyl)ation of p53 contributes to TPEN-induced neuronal apoptosis.: This study investigates the role of poly(ADP-ribosyl)ation of the tumor suppressor protein p53 in neuronal apoptosis induced by TPEN, highlighting mechanisms of cell death in neurodegenerative diseases (Kim HL et al., 2015).

  • Essential role of p53 in TPEN-induced neuronal apoptosis.: This article explores the critical involvement of the p53 protein in apoptosis initiated by TPEN in neuronal cells, offering insights into potential therapeutic targets for neuroprotection (Ra H et al., 2009).

  • Clioquinol induces autophagy in cultured astrocytes and neurons by acting as a zinc ionophore.: Demonstrates how Clioquinol, similar to TPEN, functions as a zinc ionophore to induce autophagy, particularly in neurological contexts, which may be relevant for understanding TPEN′s mechanisms (Park MH et al., 2011).

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C) under an inert gas. Alternately, acidify (pH ≤3) with HCl for long term storage. Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Fonte, J., et al. 2001. J. Alzheimer’s Dis. 3, 209.
Cherny, R.A., et al. 1999. J. Biol. Chem. 274, 23223.
Aballay, A., et al. 1995. Biochem. J.312, 919.
Jiang, S., et al. 1995. Lab. Invest.73, 111.
Sheridan, R.E. and Deshpande, S.S. 1995. Toxicon33, 539.
McCabe, M.J., et al. 1993. Lab. Invest. 69, 101.
Hinkle, P.M., et al. 1992. J. Biol. Chem.267, 25553.
Baba, A., et al. 1991. Brain Res. 557, 103.
Forbes, I.J., et al. 1991. Exp. Cell Res. 195, 224.
Csermely, P. and Somogyi, J. 1989. J. Cellular Physiol.138, 593.
Raspe, E., et al. 1989. Eur. J. Pharmacol.163, 345.
Arslan, L.P., et al. 1985. J. Biol. Chem.260, 2719.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service