Skip to Content
Merck
All Photos(1)

Key Documents

614850

Sigma-Aldrich

Topoisomerase I, His•Tag®, Human, Wild Type, Recombinant, S. frugiperda

Synonym(s):

Topo I, Wild Type

Sign Into View Organizational & Contract Pricing

Select a Size

1 SET
€1,060.00

€1,060.00


Estimated to ship on29 April 2025Details



Select a Size

Change View
1 SET
€1,060.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.54

€1,060.00


Estimated to ship on29 April 2025Details


recombinant

expressed in insect cells (S. frugiperda)

Quality Level

Assay

≥95% (SDS-PAGE)

form

liquid

specific activity

1 units/ng protein

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

shipped in

wet ice

storage temp.

−70°C

General description

Recombinant, human topoisomerase I fused to a His•Tag sequence and expressed in S. frugiperda insect cells. It catalyzes the relaxation of both positive and negative supercoiled DNAs without the requirement of energy. In addition to DNA replication and transcriptional activation, DNA topoisomerase I also plays a role in pre-mRNA splicing, cell cycle and other gene regulatory pathways during cell growth and development. DNA Topo I is an important target for ant-tumor agents as a result of the fact that is causes single-strand DNA breaks of the type that are present in most cancers.

Biochem/physiol Actions

0.1-1 unit (ng) is sufficient for relaxing 0.5 µg of pBR322 supercoiled DNA in a 20 µl reaction.

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Standard Handling (A)

Physical form

5000 U enzyme in 100 mM KCl, 20 mM Tris-HCl, 1 mM DTT, 200 µM EDTA, 20% glycerol, pH 7.9 and 1 vial dilution buffer (100 mM KCl, 20 mM Tris-HCl, 1 mM DTT, 0.2 mM EDTA, 20% glycerol, pH 8.0).

Reconstitution

Following initial thaw aliquot and freeze (-70°C). For storage of diluted protein, dilute in storage buffer with 500µg/ml molecular biology grade BSA, aliquot, and freeze (-70°C).

Other Notes

Pommier, Y., et al. 1998. Biochim. Biophys. Acta1400, 83.
Pourquier, P., et al. 1997. J. Biol. Chem.272, 26441.
Rossi, F., et al. 1996. Nature381, 80.
Sekiguchi, J., et al. 1996. Proc. Natl. Acad. Sci. USA93, 785.
Giovanella, B.C., et al. 1989. Science246, 1046.
Wang, J,C. 1985. Annu. Rev. Biochem.54, 665.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
HIS TAG is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service