Skip to Content
Merck
All Photos(1)

Key Documents

609310

Sigma-Aldrich

Thyroglobulin, Bovine Thyroid Gland

Synonym(s):

Thyroglobulin, Bovine Thyroid Gland

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
€188.00

€188.00


Estimated to ship on11 April 2025Details


Request a Bulk Order

Select a Size

Change View
100 MG
€188.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.42

€188.00


Estimated to ship on11 April 2025Details


Request a Bulk Order

biological source

bovine thyroid

Quality Level

Assay

≥90% (SDS-PAGE)

form

lyophilized solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

solubility

aqueous buffer: soluble
water: soluble

shipped in

ambient

storage temp.

−20°C

General description

Native thyroglobulin, consisting of two polypeptide homodimers of 330 kDa each, purified from bovine thyroid. This protein is an excellent source of N-linked glycans of various structures, including sialylated core fucose and oligomannose oligosaccharides. Gal-3-SO4 and GalNAc-6-SO4 residues, located in a single multi-branched complex type glycan, have been reported in calf thymus and human proteins.

Warning

Toxicity: Harmful (C)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 1 year at -20°C.

Other Notes

Rawitch, A.B., et al. 1993. Arch Biochem. Biophys. 300, 271.
Spiro, R.G. and Bhoyroo, V.D. 1988. J. Biol. Chem. 263, 14351.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service