Skip to Content
Merck
All Photos(1)

Key Documents

598226

Sigma-Aldrich

Thiostrepton

≥98% (HPLC), powder, FOXM1 inhibitor, Calbiochem®

Synonym(s):

Thiostrepton, FOXM1 Inhibitor I

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€130.00
10 G
€994.00

€130.00


Estimated to ship on14 April 2025Details



Select a Size

Change View
1 G
€130.00
10 G
€994.00

About This Item

Empirical Formula (Hill Notation):
C72H85N19O18S5
CAS Number:
Molecular Weight:
1664.89
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€130.00


Estimated to ship on14 April 2025Details


Product Name

Thiostrepton, Thiostrepton, CAS 1393-48-2, is an antibiotic that inhibits protein synthesis by preventing binding of GTP to 50S ribosomal subunit.

description

Merck USA index - 14, 9364

Quality Level

Assay

≥98% (HPLC)

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated
protect from light

color

off-white

solubility

DMSO: 10 mg/mL
chloroform: 10 mg/mL

shipped in

ambient

storage temp.

2-8°C

InChI

1S/C72H85N19O18S5/c1-14-26(3)47-63(105)78-30(7)57(99)75-28(5)56(98)76-31(8)58(100)91-72-19-18-40(66-85-43(22-111-66)59(101)77-29(6)55(97)74-27(4)54(73)96)81-52(72)42-21-112-67(83-42)49(34(11)109-69(107)41-20-37(32(9)92)36-16-17-39(79-47)51(95)50(36)80-41)89-60(102)44-24-113-68(86-44)53(71(13,108)35(12)94)90-62(104)45-23-110-65(84-45)38(15-2)82-64(106)48(33(10)93)88-61(103)46-25-114-70(72)87-46/h15-17,20-22,24-26,30-35,39,45,47-49,51-53,79,92-95,108H,4-6,14,18-19,23H2,1-3,7-13H3,(H2,73,96)(H,74,97)(H,75,99)(H,76,98)(H,77,101)(H,78,105)(H,82,106)(H,88,103)(H,89,102)(H,90,104)(H,91,100)/b38-15+

InChI key

NSFFHOGKXHRQEW-DVRIZHICSA-N

General description

A thiazole-containing peptide antibiotic that inhibits protein synthesis by preventing binding of GTP to 50S ribosomal subunit. Inhibits the function of elongation factor G (EF-G) and the dissociation of EF-G from the ribosome. The thiostrepton-resistant gene is also commonly used as a selective marker for recombinant DNA/plasmid technologies.
A thiazole-containing polypeptide antibiotic isolated from Streptomyces azureus. Inhibits bacterial protein synthesis and ribosomal GTPase activity by binding noncovalently, but virtually irreversibly, to the 23S rRNA in the GTPase center of the 50S subunit. Thiostrepton binding directly prevents binding to the ribosome of elongation factor G (EF-G), which also binds specifically to the 23S rRNA. Thiostrepton has proved useful in elucidating the molecular mechanisms of translation, particularly of ribosomal translocation. It is also used for selection, in bacteria of Streptomyces sp., of vectors containing the tsr gene (S. azureus), which encode a 23S rRNA methylase whose action prevents thiostrepton from binding to ribosomes.

Biochem/physiol Actions

Primary Target
EF-G

Warning

Toxicity: Standard Handling (A)

Other Notes

Rodnina, M.V., et al. 1999. Proc. Natl. Acad. Sci USA96, 9586.
Rosendahl, G. and Douthwaite, S. 1994. Nucleic Acids Res.22, 357.
Saarma, U. and Remme, J. 1992. Nucleic Acids Res.20, 3147.
Malina, H. and Robert-Gero, M. 1992. Appl. Environ. Microbiol.58, 895.
Blanco, G., et al. 1992. Gene112, 59.
Miller, S.P. and Bodley, J.W. 1991. Nucleic Acids Res.19, 1657.
Ryan, P.C., et al. 1991. J. Mol. Biol.221, 1257.
Kutay, U.R., et al. 1990. Biochim. Biophys. Acta1050, 193.
Smokvina, T., et al. 1990. Gene94, 53.
Garrett, R. 1983. Trends Biochem. Sci.8, 189.
Bodley, J.W., et al. 1970. Biochem. Biophys. Res. Commun.41, 1406.
Thompson, J. and Cundliffe, E. 1970. Biochimie 73, 1131.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cassie Liu et al.
Journal of ovarian research, 17(1), 94-94 (2024-05-05)
Genetic studies implicate the oncogenic transcription factor Forkhead Box M1 (FOXM1) as a potential therapeutic target in high-grade serous ovarian cancer (HGSOC). We evaluated the activity of different FOXM1 inhibitors in HGSOC cell models. We treated HGSOC and fallopian tube
Sarah Franco et al.
Heliyon, 6(6), e04028-e04028 (2020-06-25)
Accelerated smooth muscle cell (SMC) proliferation is the primary cause of intimal hyperplasia (IH) following vascular interventions. Forkhead Box M1 (FOXM1) is considered a proliferation-associated transcription factor. However, the presence and role of FOXM1 in IH following vascular injury have
Chiara Battistini et al.
Cell death & disease, 15(5), 370-370 (2024-05-29)
In ovarian tumors, the omental microenvironment profoundly influences the behavior of cancer cells and sustains the acquisition of stem-like traits, with major impacts on tumor aggressiveness and relapse. Here, we leverage a patient-derived platform of organotypic cultures to study the
Gia-Buu Tran et al.
Cancer research, 83(13), 2248-2261 (2023-04-15)
High-risk neuroblastoma exhibits transcriptional activation of the mevalonate pathway that produces cholesterol and nonsterol isoprenoids. A better understanding of how this metabolic reprogramming contributes to neuroblastoma development could help identify potential prevention and treatment strategies. Here, we report that both
Sonali Sinha et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 34(10), 13597-13608 (2020-08-20)
Acute kidney injury (AKI) is characterized by injury to the tubular epithelium that leads to the sudden loss of renal function. Proper tubular regeneration is essential to prevent progression to chronic kidney disease. In this study, we examined the role

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service