Skip to Content
Merck
All Photos(1)

Key Documents

5.89593

Sigma-Aldrich

Methyl acetate

99.5%, anhydrous

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
€77.00
1 L
€152.00

€77.00


Estimated to ship on02 October 2025Details


Request a Bulk Order

Select a Size

Change View
100 ML
€77.00
1 L
€152.00

About This Item

Linear Formula:
CH3COOCH3
CAS Number:
Molecular Weight:
74.08
Beilstein:
1736662
MDL number:
UNSPSC Code:
12352108
NACRES:
NA.22
grade:
anhydrous
bp:
57-58 °C (lit.)
vapor pressure:
165 mmHg ( 20 °C)

€77.00


Estimated to ship on02 October 2025Details


Request a Bulk Order

grade

anhydrous

Quality Level

vapor density

2.55 (vs air)

vapor pressure

165 mmHg ( 20 °C)

Assay

99.5%

autoignition temp.

936 °F

expl. lim.

16 %

refractive index

n20/D 1.361 (lit.)

bp

57-58 °C (lit.)

mp

−98 °C (lit.)

density

0.934 g/mL at 25 °C

SMILES string

COC(C)=O

InChI

1S/C3H6O2/c1-3(4)5-2/h1-2H3

InChI key

KXKVLQRXCPHEJC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

8.6 °F - closed cup

Flash Point(C)

-13 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service