Skip to Content
Merck
All Photos(1)

Key Documents

5.89589

Sigma-Aldrich

Diethyl ether

≥99.7%, anhydrous, contains 1 ppm BHT as inhibitor

Synonym(s):

Et2O, Ethoxyethane, Ether, Ethyl ether

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
€56.50
250 ML
€97.10
1 L
€118.00
2.5 L
€299.00

€56.50


Estimated to ship on31 March 2025Details


Request a Bulk Order

Select a Size

Change View
100 ML
€56.50
250 ML
€97.10
1 L
€118.00
2.5 L
€299.00

About This Item

Linear Formula:
(CH3CH2)2O
CAS Number:
Molecular Weight:
74.12
Beilstein:
1696894
MDL number:
UNSPSC Code:
12352112
NACRES:
NA.22
grade:
anhydrous
bp:
34.6 °C (lit.)

€56.50


Estimated to ship on31 March 2025Details


Request a Bulk Order

Product Name

Diethyl ether, anhydrous, ≥99.7%, contains 1 ppm BHT as inhibitor

grade

anhydrous

Quality Level

vapor density

2.6 (vs air)

Assay

≥99.7%

autoignition temp.

320 °F

contains

1 ppm BHT as inhibitor

expl. lim.

36.5 %

refractive index

n20/D 1.3530 (lit.)

bp

34.6 °C (lit.)

mp

−116 °C (lit.)

density

0.706 g/mL at 25 °C (lit.)

SMILES string

CCOCC

InChI

1S/C4H10O/c1-3-5-4-2/h3-4H2,1-2H3

InChI key

RTZKZFJDLAIYFH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Flam. Liq. 1 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

-40.0 °F - closed cup

Flash Point(C)

-40 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service