Skip to Content
Merck
All Photos(1)

Key Documents

5.89583

Sigma-Aldrich

Dibutyl ether

99.3%, anhydrous

Synonym(s):

Butyl ether

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
€75.80
1 L
€184.00
2 L
€335.00

€75.80


Estimated to ship on03 November 2025Details


Request a Bulk Order

Select a Size

Change View
100 ML
€75.80
1 L
€184.00
2 L
€335.00

About This Item

Linear Formula:
[CH3(CH2)3]2O
CAS Number:
Molecular Weight:
130.23
Beilstein:
1732752
MDL number:
UNSPSC Code:
12191502
NACRES:
NA.22
grade:
anhydrous
bp:
142-143 °C (lit.)
vapor pressure:
4.8 mmHg ( 20 °C)

€75.80


Estimated to ship on03 November 2025Details


Request a Bulk Order

grade

anhydrous

Quality Level

vapor density

4.48 (vs air)

vapor pressure

4.8 mmHg ( 20 °C)

Assay

99.3%

autoignition temp.

365 °F

expl. lim.

8.5 %

refractive index

n20/D 1.399 (lit.)

bp

142-143 °C (lit.)

mp

−98 °C (lit.)

solubility

H2O: 0.113 g/L at 20 °C

density

0.764 g/mL at 25 °C (lit.)

SMILES string

CCCCOCCCC

InChI

1S/C8H18O/c1-3-5-7-9-8-6-4-2/h3-8H2,1-2H3

InChI key

DURPTKYDGMDSBL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Classifications

Aquatic Chronic 3 - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

82.4 °F - closed cup

Flash Point(C)

28 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service