Skip to Content
Merck
All Photos(1)

Key Documents

580560

Millipore

TCEP, Hydrochloride

A water-soluble and odorless disulfide-reducing agent that is more stable and effective than DTT.

Synonym(s):

TCEP, Hydrochloride, tris(2-Carboxyethyl)phosphine, HCl

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
Inquire


Estimated to ship on27 January 2025Details


Request a Bulk Order

Select a Size

Change View
1 G
Inquire

About This Item

Empirical Formula (Hill Notation):
C9H15O6P · xHCl
CAS Number:
Molecular Weight:
250.19 (free base basis)
MDL number:
UNSPSC Code:
12352128
NACRES:
NA.25
form:
crystalline solid
Assay:
≥98% (elemental analysis)


Estimated to ship on27 January 2025Details


Request a Bulk Order

Quality Level

Assay

≥98% (elemental analysis)

form

crystalline solid

reaction suitability

reagent type: reductant

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated

color

white

solubility

water: 5 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

P(CCC(=O)[O-])(CCC(=O)[O-])CCC(=O)[O-]

InChI

1S/C9H15O6P/c10-7(11)1-4-16(5-2-8(12)13)6-3-9(14)15/h1-6H2,(H,10,11)(H,12,13)(H,14,15)/p-3

InChI key

PZBFGYYEXUXCOF-UHFFFAOYSA-K

General description

A water-soluble and odorless disulfide reducing agent which is more stable and effective than DTT. TCEP reduces even the most stable water-soluble alkyl disulfides at pH 4.5 within 5 min. TCEP is also capable of functioning as a reducing agent even in pH range of 1.5-8.5. Is more resistant to air-oxidation as compared to DTT.

Application


  • Peptide and Protein Desulfurization with Diboron Reagents.: This study investigates the use of TCEP hydrochloride in peptide and protein desulfurization, demonstrating its effectiveness in facilitating diboron-mediated reactions. (Jing & Walczak, 2024).

  • A Study on Thiol-Michael Addition to Semi-Synthetic Elastin-Hyaluronan Material for Electrospun Scaffolds.: TCEP hydrochloride is utilized to enhance the thiol-Michael addition reaction in the development of electrospun scaffolds, contributing to advanced biomaterials research. (Laezza et al., 2024).

  • Optimized total RNA isolation from bovine sperm with enhanced sperm head lysis.: This paper presents an optimized protocol for RNA isolation from bovine sperm, highlighting the role of TCEP hydrochloride in improving sperm head lysis efficiency. (Tiwari et al., 2024).

  • Target Recognition-Triggered Peroxidase-Mimicking Activity Depression in Homochiral Nanochannels for Identifying Cystine Enantiomers.: The study uses TCEP hydrochloride in a novel peroxidase-mimicking system to differentiate cystine enantiomers, showcasing its application in chiral analysis. (Xu et al., 2023).

  • In situ digestion of alcohol-fixed cells for quantitative proteomics.: TCEP hydrochloride is applied in the in situ digestion of fixed cells to enhance the accuracy of quantitative proteomics analyses. (Hatano et al., 2023).

Packaging

Packaged under inert gas

Warning

Toxicity: Corrosive (L)

Other Notes

Due to the nature of the Hazardous Materials in this shipment, additional shipping charges may be applied to your order. Certain sizes may be exempt from the additional hazardous materials shipping charges. Please contact your local sales office for more information regarding these charges.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service