Skip to Content
Merck
All Photos(2)

Key Documents

574661

Sigma-Aldrich

Survivin Antagonist, S12

The Survivin Antagonist, S12 controls the biological activity of Survivin.

Synonym(s):

Survivin Antagonist, S12, Survivin Inhibitor II, 3-(2-(benzo[d][1,3]dioxol-5-yl)-2-oxoethyl)-5-bromo-3-hydroxyindolin-2-one

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€188.00

€188.00


Estimated to ship on22 April 2025Details



Select a Size

Change View
10 MG
€188.00

About This Item

Empirical Formula (Hill Notation):
C17H12BrNO5
CAS Number:
Molecular Weight:
390.18
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€188.00


Estimated to ship on22 April 2025Details


Quality Level

Assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 50 mg/mL, nearly colorless

shipped in

ambient

storage temp.

−20°C

SMILES string

O=C(CC1(C(C=C(Br)C=C2)=C2NC1=O)O)C3=CC(OCO4)=C4C=C3

InChI

1S/C17H12BrNO5/c18-10-2-3-12-11(6-10)17(22,16(21)19-12)7-13(20)9-1-4-14-15(5-9)24-8-23-14/h1-6,22H,7-8H2,(H,19,21)

InChI key

WFYHAKOLBICHEZ-UHFFFAOYSA-N

General description

A cell-permeable hydroxyindolinone compound that acts as a selective antagonist of survivin. Induces mitotic arrest and blocks cell cycle at G2/M phase (~40 µM in HeLA cells) leading to apoptotic cell death. Shown to bind to a specific cavity adjacent to the interface involved in the dimerization of survivin. Binding is specific only to native survivin and does not affect any mutant forms. Inhibits HCT116 cell proliferation independently of their p53 status and reduces tumor volume in xenograft models in a dose-dependent manner (5 to 15 mg/kg).

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Berezov, A., et al. 2011. Oncogene31, 1938.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service