5744
NovaSeptum® SURe High Purity Bag Sampling Unit
Manifold (5-valve) with Inlet C-Flex® Tubing, sterile; β-irradiated, volume 100 mL
Synonym(s):
HP bag, High Purity bag, bag
Select a Size
€2,162.00
In StockDetails
Select a Size
About This Item
€2,162.00
In StockDetails
Recommended Products
material
C-Flex tubing
TPE tubing
polycarbonate Luer lock
polyethylene body
polyethylene container
Quality Level
sterility
sterile; β-irradiated
product line
NovaSeptum® SURe
storage condition
room temperature
parameter
-80-50 °C temp. range (-112-122 °F) (when used with a NovaSeptum® Case)
0.30 bar max. pressure (4.35 psi)
capacity
100 mL
volume
100 mL
fitting
Luer-Lok® inlet connection (3-piece)
inlet connection (containing a male, female and an injection site)
Luer-Lok® outlet connection (3-piece)
outlet connection (containing a male, female and an injection site)
application(s)
sterile sampling
Looking for similar products? Visit Product Comparison Guide
General description
This product comes as a manifolded unit with 5 high purity bags and connects via C-flex® tubing for thermowelding. The closed design ensures your sample will be isolated from point of sample to analysis, reducing the risk of losing valuable product and maintaining the integrity of your fluid samples.
Application
Features and Benefits
- Product design ensures process integrity and a representative sample from single-use manufacturing processes
- Industry-standard aseptic connection and disconnection for easy integration into your single-use process
- Other container options available: bags, bottles, syringes, and conical tubes
- Stocked and ready-to-ship for fast delivery
Packaging
Preparation Note
Analysis Note
Other Notes
Legal Information
also commonly purchased with this product
related product
required but not provided
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service