Skip to Content
Merck
All Photos(1)

Key Documents

573462

Sigma-Aldrich

Succinyl-Ala-Ala-Pro-Phe-p-nitroanilide

Sensitive chromogenic substrate for human leukocyte cathepsin G that is not reactive with elastase.

Synonym(s):

Succinyl-Ala-Ala-Pro-Phe-p-nitroanilide

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
€84.70

€84.70


Estimated to ship on08 April 2025Details


Request a Bulk Order

Select a Size

Change View
25 MG
€84.70

About This Item

Empirical Formula (Hill Notation):
C30H36N6O9
CAS Number:
Molecular Weight:
624.64
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€84.70


Estimated to ship on08 April 2025Details


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated

color

white to off-white

solubility

DMSO: 100 mg/mL

shipped in

ambient

storage temp.

−20°C

SMILES string

[N+](=O)([O-])c1ccc(cc1)N[C@@H](Cc3ccccc3)C(=O)NC(=O)[C@H]2N(CCC2)C(=O)[C@@H](NC(=O)[C@@H](NC(=O)CCC(=O)O)C)C

InChI

1S/C30H36N6O9/c1-18(31-25(37)14-15-26(38)39)27(40)32-19(2)30(43)35-16-6-9-24(35)29(42)34-28(41)23(17-20-7-4-3-5-8-20)33-21-10-12-22(13-11-21)36(44)45/h3-5,7-8,10-13,18-19,23-24,33H,6,9,14-17H2,1-2H3,(H,31,37)(H,32,40)(H,38,39)(H,34,41,42)/t18-,19-,23-,24-/m0/s1

InChI key

NZSFNPKTEBHXQS-WJNSRDFLSA-N

General description

Sensitive chromogenic substrate for human leukocyte cathepsin G that is not reactive with elastase. Also, sensitive substrate for chymotrypsin, capable of detecting nanogram amounts.

Biochem/physiol Actions

Cell permeable: no
Primary Target
Chromogenic substrate for human leukocyte cathepsin G
Product does not compete with ATP.
Reversible: no

Warning

Toxicity: Standard Handling (A)

Other Notes

Barrett, A.J. 1981. Methods Enzymol.80, 561.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service