5711
Streptomycin Sulfate, Streptomyces sp.
Synonym(s):
Streptomycin Sulfate, Streptomyces sp.
Select a Size
€86.40
Select a Size
About This Item
€86.40
Recommended Products
form
powder
Quality Level
specific activity
≥720 IU/g (dry basis)
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
color
white to off-white
shipped in
ambient
storage temp.
2-8°C
SMILES string
[S](=O)(=O)(O)O.[S](=O)(=O)(O)O.[S](=O)(=O)(O)O.N([C@@H]4[C@@H](O[C@H]([C@@H]([C@H]4O)O)CO)O[C@H]5[C@@H](O[C@H]([C@]5(O)C=O)C)O[C@@H]6[C@H]([C@@H]([C@H]([C@@H]([C@H]6O)O)N=C(N)N)O)N=C(N)N)C.N([C@@H]1[C@@H](O[C@H]([C@@H]([C@H]1O)O)CO)O[C@H]2[C@@H](O[C@H]([
InChI
1S/2C21H39N7O12.3H2O4S/c2*1-5-21(36,4-30)16(40-17-9(26-2)13(34)10(31)6(3-29)38-17)18(37-5)39-15-8(28-20(24)25)11(32)7(27-19(22)23)12(33)14(15)35;3*1-5(2,3)4/h2*4-18,26,29,31-36H,3H2,1-2H3,(H4,22,23,27)(H4,24,25,28);3*(H2,1,2,3,4)/t2*5-,6-,7+,8-,9-,10-,11+,12-,13-,14+,15+,16-,17-,18-,21+;;;/m00.../s1
InChI key
QTENRWWVYAAPBI-YCRXJPFRSA-N
General description
Biochem/physiol Actions
initiation, elongation, and termination of protein synthesis in prokaryotes and induces misreading of the genetic code
Warning
Preparation Note
Reconstitution
Other Notes
Maniatis, T., et al. 1989. In Molecular Cloning, A Laboratory Manual, Second Edition. Cold Spring Harbor, New York, p. A.6.
Modolell, J. and Davis, B.D. 1969. Nature224, 345.
Legal Information
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Repr. 2
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service