Skip to Content
Merck
All Photos(1)

Documents

569406

Sigma-Aldrich

Stearoyl-CoA Desaturase 1 Inhibitor, MF-438

Stearoyl-CoA Desaturase 1 Inhibitor, MF-438, CAS 921605-87-0, is a cell-permeable inhibitor of Stearoyl-CoA Desaturase 1 (SCD1; IC50 of 2.3 nM).

Synonym(s):

Stearoyl-CoA Desaturase 1 Inhibitor, MF-438, 2-methyl-5-(6-(4-(2-(trifluoromethyl)phenoxy)piperidin-1-yl)pyridazin-3-yl)-1,3,4-thiadiazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H18F3N5OS
Molecular Weight:
421.44
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥99% (HPLC)

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

off-white

solubility

DMSO: 10 mg/mL

shipped in

ambient

storage temp.

2-8°C

General description

A cell-permeable, thiadiazole-pyridazine compound that acts as an inhibitor of Stearoyl-CoA Desaturase 1 (SCD1) with an IC50 of 2.3 nM. In vivo, it exhibits an ED50 of 1-3 mg/kg, p.o., in mouse liver PD assays.

Warning

Toxicity: Standard Handling (A)

Other Notes

Leger, S., et al. 2010. Bioorg Med Chem Lett.2, 499.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service