Skip to Content
Merck
All Photos(1)

Key Documents

569393

Sigma-Aldrich

Stauprimide

A cell-permeable ESCs (Embryonic Stem Cells) EMT (Epithelial-Mesenchymal Transition) inducer that is reported to prime/sensitize human and murine ESC cultures for much more enhanced/efficient differentiation into progenitor cells of multiple lineages.

Synonym(s):

Stauprimide, ESC EMT Inducer, Spd

Sign Into View Organizational & Contract Pricing

Select a Size

500 μG
€233.00

€233.00


Estimated to ship on17 April 2025Details


Request a Bulk Order

Select a Size

Change View
500 μG
€233.00

About This Item

Empirical Formula (Hill Notation):
C35H28N4O5
CAS Number:
Molecular Weight:
584.62
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€233.00


Estimated to ship on17 April 2025Details


Request a Bulk Order

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

yellow

solubility

ethanol: 10 mg/mL
DMSO: 25 mg/mL

shipped in

wet ice

storage temp.

−20°C

SMILES string

[n]21c3c4[n](c7c(c4c8c(c3c9c2cccc9)[c]([nH][c]8=O)=O)cccc7)[C@@H]5O[C@]1([C@@H]([C@@H](C5)N(C)C(=O)c6ccccc6)OC)C

InChI

1S/C35H28N4O5/c1-35-31(43-3)23(37(2)34(42)18-11-5-4-6-12-18)17-24(44-35)38-21-15-9-7-13-19(21)25-27-28(33(41)36-32(27)40)26-20-14-8-10-16-22(20)39(35)30(26)29(25)38/h4-16,23-24,31H,17H2,1-3H3,(H,36,40,41)/t23-,24-,31-,35+/m1/s1

InChI key

MQCCJEYZKWZQHU-MPRCCEKMSA-N

General description

A cell-permeable ESCs (Embryonic Stem Cells) EMT (Epithelial-Mesenchymal Transition) inducer that is reported to prime/sensitize human and murine ESC cultures for much more enhanced/efficient differentiation into progenitor cells of multiple lineages, including Activin A- (Cat. No. 114700) induced definitive endoderm differentiation (60% differentiation from murine R1 ESCs with 5 ng/ml Activin A and 200 nM Spd), ATRA- (Cat. No. 554720) induced neuronal progenitor differentiation, and BMP-4- (Cat. No. 203642) induced mesoderm differentiation, under suboptimal lineage specifying stimuli concentrations and often serum-free conditions, while Spd sensitization alone does not induce any ESCs differentiation. Spd is shown to interact with NME2/DNPK B/PUF and prevent NME2 nuclear localization and c-myc transcription in murine R1 ESC cultures (46% and 85% drop in c-myc mRNA in 24 h, respectively, with 200 nM and 500 nM Spd), which accounts for, at least in part, its EMT induction activity. Although a structural analog of the broad-spectrum kinase inhibitors Staurosporine (Cat. Nos. 569396 and 569397) and UCN-01 (Cat. No. 539644), Spd exhibits significant kinase inhibitory activities only at elevated concentrations, but not at concentrations ≤500 nM typically used in EMT induction.

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliqout and freeze (-70°C). Stock solutions are stable for up to 6 months at -70°C.

Other Notes

Zhu, S., et al. 2009. Cell Stem Cell4, 416.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service